Bathocuproin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline

140910-1G 140910-500MG
SIAL140910-1GEA 131 EUR
SIAL140910-1G SIAL140910-500MG
Bathocuproin, Sigma-Aldrich®
Bathocuproin
Formel: C₂₆H₂₀N₂
Molecular Weight: 360,46 g/mol
MDL: MFCD00004972
CAS-Nummer: 4733-39-5
EINECS: 225-240-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR