1-(2-Piperidinoethyl)-1H-pyrazol

Lieferant: COMBI-BLOCKS

HI-1527-5G HI-1527-1G HI-1527-250M
COBBHI-1527-5GEA 1810 EUR
COBBHI-1527-5G COBBHI-1527-1G COBBHI-1527-250M
1-(2-Piperidinoethyl)-1H-pyrazol
1-(2-Piperidinoethyl)-1H-pyrazol
Formel: C₁₀H₁₇N₃
MDL: MFCD03727253
CAS-Nummer: 1187385-66-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR