4-Brom-3,5-dimethyl-1-(2-tetrahydropyranyl)-1H-pyrazol
Lieferant: COMBI-BLOCKS
HI-1570-5G
HI-1570-1G
COBBHI-1570-5GEA
739
EUR
COBBHI-1570-5G
COBBHI-1570-1G
4-Brom-3,5-dimethyl-1-(2-tetrahydropyranyl)-1H-pyrazol
4-Brom-3,5-dimethyl-1-(2-tetrahydropyranyl)-1H-pyrazol
Formel:
C₁₀H₁₅BrN₂O Molecular Weight: 259,15 g/mol |
MDL:
MFCD11844792 CAS-Nummer: 1187385-69-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...