1-(1-Boc-piperidino)-1H-pyrazol-4-boronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: Crizotinib Impurity 5, 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]piperidine-1-carboxylic acid tert-Butyl ester, tert-Butyl 4-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)piperidine-1-carboxylate, 1-(1-Boc-piperidin-4-yl)pyrazole-4-boronic acid Pinacol ester, 2-[1-(1-Boc-piperidin-4-yl)pyrazol-4-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, tert-Butyl 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]piperidine-1-carboxylate

FM-2957-1G FM-2957-250M
COBBFM-2957-1GEA 25.9 EUR
COBBFM-2957-1G COBBFM-2957-250M
1-(1-Boc-piperidino)-1H-pyrazol-4-boronsäurepinakolester
1-(1-Boc-piperidino)-1H-pyrazol-4-boronsäurepinakolester
Formel: C₁₉H₃₂BN₃O₄
Molecular Weight: 377.2861 g/mol
MDL: MFCD11112131
CAS-Nummer: 877399-74-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR