1-(1-Boc-piperidino)-1H-pyrazol-4-boronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (1-(1-(tert-Butoxycarbonyl)piperidin-4-yl)-1H-pyrazol-4-yl)boronic acid

BB-6007-1G BB-6007-250M
COBBBB-6007-1GEA 0 EUR
COBBBB-6007-1G COBBBB-6007-250M
1-(1-Boc-piperidino)-1H-pyrazol-4-boronsäure
1-(1-Boc-piperidino)-1H-pyrazol-4-boronsäure
Formel: C₁₃H₂₂BN₃O₄
MDL: MFCD12913942
CAS-Nummer: 1190875-39-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR