1-(1-Boc-piperidino)-4-iod-1H-pyrazol

Lieferant: COMBI-BLOCKS

Synonyme: 1-Boc-4-(4-Iodo-1H-pyrazol-1-yl)piperidine, 1-(1-Boc-4-piperidinyl)-4-iodopyrazole

AM-1738-5G AM-1738-1G
COBBAM-1738-5GEA 798 EUR
COBBAM-1738-5G COBBAM-1738-1G
1-(1-Boc-piperidino)-4-iod-1H-pyrazol
1-(1-Boc-piperidino)-4-iod-1H-pyrazol
Formel: C₁₃H₂₀IN₃O₂
Molecular Weight: 377,23 g/mol
MDL: MFCD12913927
CAS-Nummer: 877399-73-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR