5-Brom-3-nitro-2-(1H-pyrazol-1-yl)pyridin

Lieferant: COMBI-BLOCKS

PY-7159-1G PY-7159-5G
COBBPY-7159-1GEA 98.3 EUR
COBBPY-7159-1G COBBPY-7159-5G
5-Brom-3-nitro-2-(1H-pyrazol-1-yl)pyridin
5-Brom-3-nitro-2-(1H-pyrazol-1-yl)pyridin
Formel: C₈H₅BrN₄O₂
MDL: MFCD09801050
CAS-Nummer: 446284-40-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR