2-(1H-Pyrazol-3-yl)pyridin

Lieferant: CHEMPUR

SEW04056.250 SEW04056.1
CHMPSEW04056.250EA 0 EUR
CHMPSEW04056.250 MAYBSEW04056.1
2-(1H-Pyrazol-3-yl)pyridin
2-(1H-Pyrazol-3-yl)pyridin
Formel: C₈H₇N₃
Molecular Weight: 145,16 g/mol
MDL: MFCD00115151
CAS-Nummer: 75415-03-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR