4-(1H-Pyrazol-4-yl)pyridin
Lieferant: FLUOROCHEM
016084-5G
016084-1G
FLUO016084-5GEA
1030
EUR
FLUO016084-5G
FLUO016084-1G
4-(1H-Pyrazol-4-yl)pyridin
4-(1H-Pyrazol-4-yl)pyridin
Formel:
C₈H₇N₃ Molecular Weight: 145,1613 g/mol |
MDL:
MFCD00480339 CAS-Nummer: 19959-71-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...