3-Brom-6-piperazino-2-picolin Hydrochlorid

Lieferant: COMBI-BLOCKS

Synonyme: 1-(5-Bromo-6-methylpyridin-2-yl)piperazine hydrochloride

PY-1173-5G PY-1173-1G
COBBPY-1173-5GEA 460 EUR
COBBPY-1173-5G COBBPY-1173-1G
3-Brom-6-piperazino-2-picolin Hydrochlorid
3-Brom-6-piperazino-2-picolin Hydrochlorid
Formel: C₁₀H₁₅BrClN₃
Molecular Weight: 292.6000 g/mol
MDL: MFCD12913993
CAS-Nummer: 1187386-04-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR