Ethyl-4-chlor-2-piperidinopyrimidin-5-carboxylat

Lieferant: COMBI-BLOCKS

HC-2021-1G HC-2021-250M
COBBHC-2021-1GEA 864 EUR
COBBHC-2021-1G COBBHC-2021-250M
Ethyl-4-chlor-2-piperidinopyrimidin-5-carboxylat
Ethyl-4-chlor-2-piperidinopyrimidin-5-carboxylat
Formel: C₁₂H₁₆ClN₃O₂
MDL: MFCD11111793
CAS-Nummer: 34750-24-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR