Methyl-4-cyclopropyl-2-piperidinopyrimidin-5-carboxylat

Lieferant: COMBI-BLOCKS

HC-2123-5G HC-2123-1G
COBBHC-2123-5GEA 595 EUR
COBBHC-2123-5G COBBHC-2123-1G
Methyl-4-cyclopropyl-2-piperidinopyrimidin-5-carboxylat
Methyl-4-cyclopropyl-2-piperidinopyrimidin-5-carboxylat
Formel: C₁₄H₁₉N₃O₂
MDL: MFCD11504897
CAS-Nummer: 1072944-55-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR