2-(4-Morpholino)-5-pyrimidinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-yl)morpholine, 2-(4-Morpholino)pyrimidine-5-boronic acid, pinacol eser, 2-(4-Morpholino)pyrimidine-5-boronic acid, pinacol ester

PN-8707-1G PN-8707-5G
COBBPN-8707-1GEA 181 EUR
COBBPN-8707-1G COBBPN-8707-5G
2-(4-Morpholino)-5-pyrimidinboronsäurepinakolester
2-(4-Morpholino)-5-pyrimidinboronsäurepinakolester
Formel: C₁₄H₂₂BN₃O₃
Molecular Weight: 291,1538 g/mol
MDL: MFCD07368246
CAS-Nummer: 957198-30-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR