2-(4-Boc-piperazino)-5-pyrimidinboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
tert-Butyl 4-(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-yl)piperazine-1-carboxylate
PN-8722-5G
PN-8722-1G
COBBPN-8722-5GEA
912
EUR
COBBPN-8722-5G
COBBPN-8722-1G
2-(4-Boc-piperazino)-5-pyrimidinboronsäurepinakolester
2-(4-Boc-piperazino)-5-pyrimidinboronsäurepinakolester
Formel:
C₁₉H₃₁BN₄O₄ Molecular Weight: 390.2848 g/mol |
MDL:
MFCD07781250 CAS-Nummer: 940284-98-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...