5-(1-Piperidinylmethyl)thiophen-2-boronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
1-{[5-(Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methyl}piperidine
PN-9134-5G
PN-9134-1G
COBBPN-9134-5GEA
570
EUR
COBBPN-9134-5G
COBBPN-9134-1G
5-(1-Piperidinylmethyl)thiophen-2-boronsäurepinakolester
5-(1-Piperidinylmethyl)thiophen-2-boronsäurepinakolester
Formel:
C₁₆H₂₆BNO₂S Molecular Weight: 307,26 g/mol |
MDL:
MFCD11113036 CAS-Nummer: 1218790-44-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...