2,2'-Thenil ≥98%
Lieferant: Thermo Scientific
Synonyme:
1,2-Di(thiophen-2-yl)ethane-1,2-dione
B24753.03
B24753.06
B24753.03EA
43.2
EUR
B24753.03
B24753.06
2,2'-Thenil ≥98%
2,2'-Thenil
Purity: 98%
CAS No.: 7333-07-5
Molecular Formula: C10H6O2S2
Formula Weight: 222.29
Melting Point: 81-84°
MDL No.: MFCD00173678
Formel:
C₁₀H₆O₂S₂ Molecular Weight: 222,29 g/mol Schmelzpunkt: 81…84 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173678 CAS-Nummer: 7333-07-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...