Ferrozine mono-sodium salt hydrate, Supelco®

Lieferant: Merck

Synonyme: 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazin-4,4''-disulfonsäure Mononatriumsalz Hydrat

M-Clarity™ Qualitätsstufe = MQ100

82950-1G 82950-5G
82950-1GEA 58.4 EUR
82950-1G 82950-5G
Ferrozine mono-sodium salt hydrate, Supelco®
Ferrozine mono-sodium salt hydrate
Formel: C₂₀H₁₃N₄NaO₆S₂·H₂O
MDL: MFCD00078417
CAS-Nummer: 63451-29-6
EINECS: 274-196-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=827c458da2852cd8c20c3ba00af19dbb3823ba8c&vid=827c458da2852cd8c20c3ba00af19dbb3823ba8c&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR