1-(2-Bromphenylsulfonyl)pyrrolidin
Lieferant: COMBI-BLOCKS
Synonyme:
1-((2-Bromophenyl)sulphonyl)pyrrolidine
OT-0504-1G
OT-0504-5G
COBBOT-0504-1GEA
142
EUR
COBBOT-0504-1G
COBBOT-0504-5G
1-(2-Bromphenylsulfonyl)pyrrolidin
1-(2-Bromphenylsulfonyl)pyrrolidin
Formel:
C₁₀H₁₂BrNO₂S Molecular Weight: 290,180 g/mol |
MDL:
MFCD09258650 CAS-Nummer: 929000-58-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...