1-(3-Brom-4-methylphenylsulfonyl)pyrrolidin

Lieferant: COMBI-BLOCKS

OT-0169-5G OT-0169-1G
COBBOT-0169-5GEA 303 EUR
COBBOT-0169-5G COBBOT-0169-1G
1-(3-Brom-4-methylphenylsulfonyl)pyrrolidin
1-(3-Brom-4-methylphenylsulfonyl)pyrrolidin
Formel: C₁₁H₁₄BrNO₂S
Molecular Weight: 304,2034 g/mol
MDL: MFCD07363840
CAS-Nummer: 850429-75-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR