1-(4-Bromphenylsulfonyl)pyrrolidin

Lieferant: COMBI-BLOCKS

Synonyme: 4-Bromo-N-pyrrolidin-1-ylbenzenesulphonamide, 1-((4-Bromophenyl)sulphonyl)pyrrolidine

OT-0090-5G OT-0090-1G
COBBOT-0090-5GEA 302 EUR
COBBOT-0090-5G COBBOT-0090-1G
1-(4-Bromphenylsulfonyl)pyrrolidin
1-(4-Bromphenylsulfonyl)pyrrolidin
Formel: C₁₀H₁₂BrNO₂S
Molecular Weight: 290.1768 g/mol
MDL: MFCD00584577
CAS-Nummer: 136350-52-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR