1-(4-Chlorphenylsulfonyl)pyrrolidin-2-carbonsäure

Lieferant: BIONET RESEARCH

10G-428S.500 10G-428S.1 10G-428S.5 10G-428S.10
BIOT10G-428S.500EA 0 EUR
BIOT10G-428S.500 BIOT10G-428S.1 BIOT10G-428S.5 BIOT10G-428S.10
1-(4-Chlorphenylsulfonyl)pyrrolidin-2-carbonsäure
1-(4-Chlorphenylsulfonyl)pyrrolidin-2-carbonsäure
Formel: C₁₁H₁₂ClNO₄S
MDL: MFCD00172806
CAS-Nummer: 73096-27-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR