1-Boc-isonipecotinsäureethylester, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Ethyl N-BOC-4-piperidinecarboxylate, Ethyl N-Boc-piperidine-4-carboxylate
M-Clarity™ Qualitätsstufe = MQ100
665150-25G
665150-5G
SIAL665150-25GEA
116
EUR
SIAL665150-25G
SIAL665150-5G
1-Boc-isonipecotinsäureethylester, Sigma-Aldrich®
1-Boc-isonipecotinsäureethylester
Formel:
C₁₃H₂₃NO₄ Molecular Weight: 257.33 g/mol |
MDL:
MFCD01763998 CAS-Nummer: 142851-03-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...