1-Boc-isonipecotinsäuremethylester

Lieferant: FLUOROCHEM

Synonyme: Methyl piperidine-4-carboxylate, N-BOC protected, 1-tert-Butyl 4-methyl piperidine-1,4-dicarboxylate

011534-5G 011534-1G
FLUO011534-5GEA 29.9 EUR
FLUO011534-5G FLUO011534-1G
1-Boc-isonipecotinsäuremethylester
1-Boc-isonipecotinsäuremethylester
Formel: C₁₂H₂₁NO₄
Molecular Weight: 243.2994 g/mol
MDL: MFCD02183584
CAS-Nummer: 124443-68-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR