Isonipecotamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Piperidine-4-carboxamide

M-Clarity™ Qualitätsstufe = MQ100

I17907-100G I17907-25G
SIALI17907-100GEA 104 EUR
SIALI17907-100G SIALI17907-25G
Isonipecotamid, Sigma-Aldrich®
Isonipecotamid
Formel: C₆H₁₂N₂O
Molecular Weight: 128.18 g/mol
MDL: MFCD00038012
CAS-Nummer: 39546-32-2
EINECS: 254-501-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR