Methyl-3-piperidincarboxylat Hydrochlorid
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl piperidine-3-carboxylate hydrochloride
AM-1709-25G
AM-1709-5G
COBBAM-1709-25GEA
71
EUR
COBBAM-1709-25G
COBBAM-1709-5G
Methyl-3-piperidincarboxylat Hydrochlorid
Methyl-3-piperidincarboxylat Hydrochlorid
Formel:
C₇H₁₄ClNO₂ Molecular Weight: 179.6446 g/mol |
MDL:
MFCD01861791 CAS-Nummer: 89895-55-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...