Ethyl-4-piperidincarboxylat Hydrochlorid
Lieferant: COMBI-BLOCKS
Synonyme:
Ethyl piperidine-4-carboxylate hydrochloride
OT-0794-100G
OT-0794-5G
OT-0794-25G
COBBOT-0794-100GEA
102
EUR
COBBOT-0794-100G
COBBOT-0794-5G
COBBOT-0794-25G
Ethyl-4-piperidincarboxylat Hydrochlorid
Ethyl-4-piperidincarboxylat Hydrochlorid
Formel:
C₈H₁₆ClNO₂ Molecular Weight: 193,6711 g/mol |
MDL:
MFCD06655993 CAS-Nummer: 147636-76-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...