Methyl-4-piperidincarboxylat Hydrochlorid
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl piperidine-4-carboxylate hydrochloride
AM-1021-25G
AM-1021-100G
COBBAM-1021-25GEA
53.4
EUR
COBBAM-1021-25G
COBBAM-1021-100G
Methyl-4-piperidincarboxylat Hydrochlorid
Methyl-4-piperidincarboxylat Hydrochlorid
Formel:
C₇H₁₄ClNO₂ Molecular Weight: 179.6446 g/mol |
MDL:
MFCD02667641 CAS-Nummer: 7462-86-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...