1-(Methylsulfonyl)-4-piperidincarbonsäure
Lieferant: BIONET RESEARCH
Synonyme:
1-(Methylsulphonyl)piperidine-4-carboxylic acid
3Y-0704.1
3Y-0704.10
3Y-0704.5
3Y-0704.500
BIOT3Y-0704.1EA
0
EUR
BIOT3Y-0704.1
BIOT3Y-0704.10
BIOT3Y-0704.5
BIOT3Y-0704.500
1-(Methylsulfonyl)-4-piperidincarbonsäure
1-(Methylsulfonyl)-4-piperidincarbonsäure
Formel:
C₇H₁₃NO₄S Molecular Weight: 207,2500 g/mol |
MDL:
MFCD03821886 CAS-Nummer: 280772-00-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...