1-(4-Fluorphenylsulfonyl)pyrrolidin-2-carbonsäure
Lieferant: BIONET RESEARCH
10G-430S.500
10G-430S.1
10G-430S.5
10G-430S.10
BIOT10G-430S.500EA
0
EUR
BIOT10G-430S.500
BIOT10G-430S.1
BIOT10G-430S.5
BIOT10G-430S.10
1-(4-Fluorphenylsulfonyl)pyrrolidin-2-carbonsäure
1-(4-Fluorphenylsulfonyl)pyrrolidin-2-carbonsäure
Formel:
C₁₁H₁₂FNO₄S |
MDL:
MFCD00172808 CAS-Nummer: 251096-96-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...