(R)-(-)-3-Fluorpyrrolidin Hydrochlorid

Lieferant: FLUOROCHEM

Synonyme: (R)-(-)-3-Fluoropyrrolidine Hydrochloride

013156-1G 013156-500MG
FLUO013156-1GEA 97.2 EUR
FLUO013156-1G FLUO013156-500MG
(R)-(-)-3-Fluorpyrrolidin Hydrochlorid
(R)-(-)-3-Fluorpyrrolidin Hydrochlorid
Formel: C₄H₉ClFN
Molecular Weight: 125,57 g/mol
MDL: MFCD04038717
CAS-Nummer: 136725-55-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR