(R)-(-)-3-Fluorpyrrolidin Hydrochlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(R)-(-)-3-Fluoropyrrolidine Hydrochloride
M-Clarity™ Qualitätsstufe = MQ100
637505-1G
637505-5G
SIAL637505-1GEA
105
EUR
SIAL637505-1G
SIAL637505-5G
(R)-(-)-3-Fluorpyrrolidin Hydrochlorid, Sigma-Aldrich®
(R)-(-)-3-Fluorpyrrolidin Hydrochlorid
Formel:
C₄H₉ClFN Molecular Weight: 125,57 g/mol |
MDL:
MFCD04038717 CAS-Nummer: 136725-55-8 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...