(S)-(+)-3-Fluorpyrrolidin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-3-Fluoropyrrolidine hydrochloride

M-Clarity™ Qualitätsstufe = MQ100

637513-1G 637513-5G
SIAL637513-1GEA 105 EUR
SIAL637513-1G SIAL637513-5G
(S)-(+)-3-Fluorpyrrolidin Hydrochlorid, Sigma-Aldrich®
(S)-(+)-3-Fluorpyrrolidin Hydrochlorid
Formel: C₄H₉ClFN
Molecular Weight: 125,57 g/mol
MDL: MFCD04038718
CAS-Nummer: 136725-53-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR