4,4-Difluorpiperidin Hydrochlorid

Lieferant: FLUOROCHEM

Synonyme: 4,4-Difluoropiperidine HCl

009632-1G 009632.25G 009632-5G 009632-25G
FLUO009632-1GEA 45.9 EUR
FLUO009632-1G FLUO009632.25G FLUO009632-5G FLUO009632-25G
4,4-Difluorpiperidin Hydrochlorid
4,4-Difluorpiperidin Hydrochlorid
Formel: C₅H₁₀ClF₂N
Molecular Weight: 157,59 g/mol
MDL: MFCD03095381
CAS-Nummer: 144230-52-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR