1-(4-Fluorphenylsulfonyl)pyrrolidin
Lieferant: COMBI-BLOCKS
OT-0453-1G
OT-0453-5G
OT-0453-25G
COBBOT-0453-1GEA
98.3
EUR
COBBOT-0453-1G
COBBOT-0453-5G
COBBOT-0453-25G
1-(4-Fluorphenylsulfonyl)pyrrolidin
1-(4-Fluorphenylsulfonyl)pyrrolidin
Formel:
C₁₀H₁₂FNO₂S |
MDL:
MFCD00783756 CAS-Nummer: 157187-14-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...