(S)-(+)-3-Aminopiperidin Dihydrochlorid

Lieferant: COMBI-BLOCKS

Synonyme: (S)-(+)-3-Piperidinamine dihydrochloride, (S)-Piperidin-3-amine dihydrochloride

AM-1780-5G AM-1780-1G
COBBAM-1780-5GEA 122 EUR
COBBAM-1780-5G COBBAM-1780-1G
(S)-(+)-3-Aminopiperidin Dihydrochlorid
(S)-(+)-3-Aminopiperidin Dihydrochlorid
Formel: C₅H₁₄Cl₂N₂
Molecular Weight: 173,08 g/mol
MDL: MFCD03427036
CAS-Nummer: 334618-07-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR