4-Methoxypiperidin

Lieferant: FLUOROCHEM

029227-1G 029227-5G
FLUO029227-1GEA 31.1 EUR
FLUO029227-1G FLUO029227-5G
4-Methoxypiperidin
4-Methoxypiperidin
Formel: C₆H₁₄ClNO
Molecular Weight: 115,1700 g/mol
MDL: MFCD04115010
CAS-Nummer: 4045-24-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR