1-Boc-piperidin-4-ylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)piperidine-1-carboxylate, Piperidine-4-boronic acid, pinacol ester, N-BOC protected

PN-9093-5G PN-9093-1G
COBBPN-9093-5GEA 269 EUR
COBBPN-9093-5G COBBPN-9093-1G
1-Boc-piperidin-4-ylboronsäurepinakolester
1-Boc-piperidin-4-ylboronsäurepinakolester
Formel: C₁₆H₃₀BNO₄
Molecular Weight: 311,2247 g/mol
MDL: MFCD09997792
CAS-Nummer: 1048970-17-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR