4-Chlor-1-methylpiperidin
Lieferant: COMBI-BLOCKS
OS-7644-25G
OS-7644-100G
COBBOS-7644-25GEA
37.5
EUR
COBBOS-7644-25G
COBBOS-7644-100G
4-Chlor-1-methylpiperidin
4-Chlor-1-methylpiperidin
Formel:
C₆H₁₂ClN Molecular Weight: 133,6192 g/mol |
MDL:
MFCD00044489 CAS-Nummer: 5570-77-4 EINECS: 226-942-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...