1-(Cyanacetyl)piperidin
Lieferant: Thermo Scientific
Synonyme:
3-Oxo-3-(piperidin-1-yl)propanenitrile
TL00706.1
TL00706EA
TL00706.50
MAYBTL00706.1EA
0
EUR
MAYBTL00706.1
MAYBTL00706EA
MAYBTL00706.50
1-(Cyanacetyl)piperidin
1-(Cyanacetyl)piperidin
Formel:
C₈H₁₂N₂O Molecular Weight: 152,20 g/mol |
MDL:
MFCD00006487 CAS-Nummer: 15029-30-8 EINECS: 239-111-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...