1-Boc-3-cyanpiperidin
Lieferant: FLUOROCHEM
Synonyme:
tert-butyl 3-cyanopiperidine-1-carboxylate, 1-Boc-3-cyanopiperidine, 3-Cyano-1-piperidinecarboxylic acid tert-Butyl ester, tert-Butyl 3-Cyano-1-piperidinecarboxylate, 1-Boc-3-piperidinecarbonitrile, 1-tert-Butoxycarbonyl-3-piperidinecarbonitrile, 1-tert-Butoxycarbonyl-3-cyanopiperidine
011523-5G
011523-1G
FLUO011523-5GEA
63.9
EUR
FLUO011523-5G
FLUO011523-1G
1-Boc-3-cyanpiperidin
1-Boc-3-cyanpiperidin
Formel:
C₁₁H₁₈N₂O₂ Molecular Weight: 210,27 g/mol |
MDL:
MFCD01861222 CAS-Nummer: 91419-53-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...