1-Boc-4-cyanpiperidin

Lieferant: FLUOROCHEM

Synonyme: tert-Butyl 4-cyanopiperidine-1-carboxylate, N-BOC-Piperidine-4-carbonitrile

011530-5G 011530-1G
FLUO011530-5GEA 60.9 EUR
FLUO011530-5G FLUO011530-1G
1-Boc-4-cyanpiperidin
1-Boc-4-cyanpiperidin
Formel: C₁₁H₁₈N₂O₂
Molecular Weight: 210.27 g/mol
MDL: MFCD01861223
CAS-Nummer: 91419-52-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR