(R)-1-Boc-piperidin-3-carbonsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(R)-1-(tert-Butoxycarbonyl)piperidine-3-carboxylic acid, Boc-D-Nip-OH, 1-Boc-D-nipecotic acid, (R)-N-BOC-piperidine-3-carboxylic acid
H52416.06
H52416.14
H52416.06EA
108
EUR
H52416.06
H52416.14
(R)-1-Boc-piperidin-3-carbonsäure ≥97%
(R)-1-Boc-piperidin-3-carbonsäure
Purity: 97%
CAS No.: 163438-09-3
Molecular Formula: C11H19NO4
Formula Weight: 229.28
Beilstein Reference: 5539297
MDL No.: MFCD00673775
Formel:
C₁₁H₁₉NO₄ Molecular Weight: 229.28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00673775 CAS-Nummer: 163438-09-3 |