(S)-1-Boc-piperidin-3-carbonsäure
Lieferant: FLUOROCHEM
Synonyme:
(S)-Boc-nipecotic acid, 1-Boc-L-nipecotic acid, (S)-1-(tert-Butoxycarbonyl)piperidine-3-carboxylic acid
019199-2G
019199-10G
FLUO019199-2GEA
0
EUR
FLUO019199-2G
FLUO019199-10G
(S)-1-Boc-piperidin-3-carbonsäure
(S)-1-Boc-piperidin-3-carbonsäure
Formel:
C₁₁H₁₉NO₄ Molecular Weight: 229.28 g/mol |
MDL:
MFCD02179172 CAS-Nummer: 88495-54-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...