1-Boc-piperidin-4-carbonsäure

Lieferant: COMBI-BLOCKS

Synonyme: 1-(tert-Butoxycarbonyl)piperidine-4-carboxylic acid, N-BOC-4-Piperidinecarboxylic acid, Boc-Inp-OH, 1-Boc-isonipecotic acid

AM-1030-5G AM-1030-25G AM-1030-100G
COBBAM-1030-5GEA 26.2 EUR
COBBAM-1030-5G COBBAM-1030-25G COBBAM-1030-100G
1-Boc-piperidin-4-carbonsäure
1-Boc-piperidin-4-carbonsäure
Formel: C₁₁H₁₉NO₄
Molecular Weight: 229,3 g/mol
MDL: MFCD00076999
CAS-Nummer: 84358-13-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR