1-Boc-piperidin-4-carbonsäure
Lieferant: FLUOROCHEM
Synonyme:
1-(tert-Butoxycarbonyl)piperidine-4-carboxylic acid, N-BOC-4-Piperidinecarboxylic acid, Boc-Inp-OH, 1-Boc-isonipecotic acid
019200-1G
019200-5G
FLUO019200-1GEA
0
EUR
FLUO019200-1G
FLUO019200-5G
1-Boc-piperidin-4-carbonsäure
1-Boc-piperidin-4-carbonsäure
Formel:
C₁₁H₁₉NO₄ Molecular Weight: 229.3 g/mol |
MDL:
MFCD00076999 CAS-Nummer: 84358-13-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...