4-(Trifluormethyl)piperidin Hydrochlorid

Lieferant: COMBI-BLOCKS

AM-2080-1G AM-2080-5G
COBBAM-2080-1GEA 43.5 EUR
COBBAM-2080-1G COBBAM-2080-5G
4-(Trifluormethyl)piperidin Hydrochlorid
4-(Trifluormethyl)piperidin Hydrochlorid
Formel: C₆H₁₁ClF₃N
Molecular Weight: 189,61 g/mol
MDL: MFCD04971993
CAS-Nummer: 155849-49-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR