4-(Trifluormethyl)piperidin Hydrochlorid

Lieferant: FLUOROCHEM

010226-1G 010226-5G
FLUO010226-1GEA 34.1 EUR
FLUO010226-1G FLUO010226-5G
4-(Trifluormethyl)piperidin Hydrochlorid
4-(Trifluormethyl)piperidin Hydrochlorid
Formel: C₆H₁₁ClF₃N
Molecular Weight: 189.61 g/mol
MDL: MFCD04971993
CAS-Nummer: 155849-49-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR