1-Boc-4-iodpiperidin

Lieferant: CHEMPUR

Synonyme: 1-BOC-4-iodo-piperidine, tert-Butyl 4-iodopiperidine-1-carboxylate

60-0171.1 60-0171.5
CHMP60-0171.1EA 0 EUR
CHMP60-0171.1 CHMP60-0171.5
1-Boc-4-iodpiperidin
1-Boc-4-iodpiperidin
Formel: C₁₀H₁₈INO₂
Molecular Weight: 311,16 g/mol
MDL: MFCD04115041
CAS-Nummer: 301673-14-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR