2-(2-(Piperidin-1-yl)ethoxy)anilin Hydrochlorid

Lieferant: COMBI-BLOCKS

HC-6930-5G HC-6930-1G
COBBHC-6930-5GEA 175 EUR
COBBHC-6930-5G COBBHC-6930-1G
2-(2-(Piperidin-1-yl)ethoxy)anilin Hydrochlorid
2-(2-(Piperidin-1-yl)ethoxy)anilin Hydrochlorid
Formel: C₁₃H₂₁ClN₂O
MDL: MFCD11053790
CAS-Nummer: 860765-11-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR