(S)-(+)-2-(Methoxymethyl)pyrrolidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-2-(Methoxymethyl)pyrrolidine

277053-500MG 277053-5G
SIAL277053-500MGEA 99.3 EUR
SIAL277053-500MG SIAL277053-5G
(S)-(+)-2-(Methoxymethyl)pyrrolidin, Sigma-Aldrich®
(S)-(+)-2-(Methoxymethyl)pyrrolidin
Formel: C₆H₁₃NO
Molecular Weight: 115,18 g/mol
MDL: MFCD00010408
CAS-Nummer: 63126-47-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR